
Trapidil impurity B

Synonym(s):
3-Amino-1,2,4-triazole, 1,2,4-Triazol-3-amine, 3-AT, Amitrol
Empirical Formula (Hill Notation):
C2H4N4
CAS Number:
Molecular Weight:
84.08
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing