
Sulfabenzamide

Synonym(s):
Sulfabenzamide, N-(4-Aminobenzenesulfonyl)benzamide, N-Sulfanilylbenzamide
Empirical Formula (Hill Notation):
C13H12N2O3S
CAS Number:
Molecular Weight:
276.31
Products
Brand
Formula Weight
Agency
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing