
Rabeprazole Related Compound F

Synonym(s):
2-{[(4-Chloro-3-methyl-2-pyridinyl)methyl]sulfinyl}-1H-benzimidazole
Empirical Formula (Hill Notation):
C14H12ClN3OS
CAS Number:
Molecular Weight:
305.78
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2