Skip to Content
Merck
Poly(ethylene glycol) methyl ether acrylate average Mn 2,000, contains MEHQ as inhibitor

Poly(ethylene glycol) methyl ether acrylate

Poly(ethylene glycol) methyl ether acrylate average Mn 2,000, contains MEHQ as inhibitor
Synonym(s):
Acryl-PEG, Methoxy PEG acrylate, Methoxy poly(ethylene glycol) monoacrylate, Poly(ethylene glycol) monomethyl ether monoacrylate, mPEG-acrylate
Linear Formula:
H2C=CHCO2(CH2CH2O)nCH3
CAS Number:

Products

Brand
Physical Form
Quality Segment
Functional Group
Functionality
Shape
Monomer Type
Polymer Composition
Polymer Type
Reaction Type
Reagent Type