Skip to Content
Merck
[Pd(allyl)Cl]2 Umicore

[Pd(allyl)Cl]2

[Pd(allyl)Cl]2 Umicore
Synonym(s):
Allylpalladium(II) chloride dimer, [PdCl(C3H5)]2
Empirical Formula (Hill Notation):
C6H10Cl2Pd2
CAS Number:
Molecular Weight:
365.89
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Reaction Type
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing