
Ondansetron impurity F

Synonym(s):
2-Methylimidazole, 2-Methylglyoxaline
Empirical Formula (Hill Notation):
C4H6N2
CAS Number:
Molecular Weight:
82.10
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing