
NECA

Synonym(s):
NECA
Empirical Formula (Hill Notation):
C12H16N6O4
CAS Number:
Molecular Weight:
308.29
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing