
Malvidin 3-glucoside chloride

Synonym(s):
Oenin chloride, Cyclamin chloride, Enin chloride, Malvidin 3-β-D-glucopyranoside, Malvidin-3-O-glucoside chloride
Empirical Formula (Hill Notation):
C23H25ClO12
CAS Number:
Molecular Weight:
528.89
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing