![GTX-6 solution 20 μg/g in acetic acid (nominal concentration), certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/252/757/f96a17a5-2028-4d2a-8e62-2e591b55fa47/640/f96a17a5-2028-4d2a-8e62-2e591b55fa47.png)
GTX-6 solution
![GTX-6 solution 20 μg/g in acetic acid (nominal concentration), certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/252/757/f96a17a5-2028-4d2a-8e62-2e591b55fa47/640/f96a17a5-2028-4d2a-8e62-2e591b55fa47.png)
Synonym(s):
(3aS,4R,10aS)-2,6-Diimino-4-({[(sulfoamino)carbonyl]oxy}methyl)-decahydropyrrolo[1,2-c]purine-5,10,10-triol solution, Gonyautoxin VI solution
Empirical Formula (Hill Notation):
C10H17N7O8S
CAS Number:
Molecular Weight:
395.35
Products
Brand
Formula Weight
Manufacturer
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-1 of 1