
Glutaryl-L-carnitine lithium salt

Synonym(s):
L-Carnitine lithium glutarate
Empirical Formula (Hill Notation):
C12H21NO6 · xLi+
CAS Number:
Molecular Weight:
275.30 (free acid basis)
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Optical Isomer
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing