
Ginkgolide B from Ginkgo biloba leaves

Synonym(s):
BN-52021
Empirical Formula (Hill Notation):
C20H24O10
CAS Number:
Molecular Weight:
424.40
Products
Product Category
Brand
Formula Weight
Purity
Format
Quality Segment
Application
Reference Material Type
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing