
Fluorescein diacetate 5-maleimide

Synonym(s):
5-Maleimido-fluorescein diacetate, Fluorescein-5-maleimide diacetate
Empirical Formula (Hill Notation):
C28H17NO9
CAS Number:
Molecular Weight:
511.44
Products
Brand
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing