
Exemestane Related Compound B

Synonym(s):
6-Hydroxymethylandrostra-1,4-diene-3,17-dione
Empirical Formula (Hill Notation):
C20H26O3
CAS Number:
Molecular Weight:
314.42
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2