
Ethyl 4-formamidobenzoate

Synonym(s):
p-Formamidobenzoic acid ethyl ester
Empirical Formula (Hill Notation):
C10H11NO3
CAS Number:
Molecular Weight:
193.20
Products
Brand
Formula Weight
Manufacturer
Product Line
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing