
Ethosuximide Impurity A

Synonym(s):
2-Ethyl-2-methylsuccinic acid
Empirical Formula (Hill Notation):
C7H12O4
CAS Number:
Molecular Weight:
160.17
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2