
Doxycycline for system suitability

Synonym(s):
Doxycycline hyclate, Doxycycline hydrochloride hemiethanolate hemihydrate
Empirical Formula (Hill Notation):
C22H24N2O8 · HCl · 0.5H2O · 0.5C2H6O
CAS Number:
Molecular Weight:
512.94
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing