
Cinnamic acid

Synonym(s):
Cinnamic acid, trans-Cinnamic acid, (2E)-3-Phenyl-2-propenoic acid, trans-3-Phenylacrylic acid
Linear Formula:
C6H5CH=CHCOOH
CAS Number:
Molecular Weight:
148.16
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing