
Cefpodoxime proxetil

Empirical Formula (Hill Notation):
C21H27N5O9S2
CAS Number:
Molecular Weight:
557.60
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing