
Boc-Glu-OBzl

Synonym(s):
Boc-L-glutamic acid 1-benzyl ester
Empirical Formula (Hill Notation):
C17H23NO6
CAS Number:
Molecular Weight:
337.37
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Quality Segment
Application
Optical Isomer
Amino Acid
Showing 1-1 of 1
Compare
Product Number
Description
Pricing