
Biotin-16-dUTP

Synonym(s):
Bio-16-dUTP tetralithium salt, 5-(N-[N-Biotinyl-ε-aminocaproyl-γ-aminobutyryl]-3-aminoallyl)-2′-deoxyuridine 5′-triphosphate
Empirical Formula (Hill Notation):
C32H52N7O18P3S
CAS Number:
Molecular Weight:
947.78
Products
Color
Formula Weight
Manufacturer
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing