
Anti-TGN46 antibody, Mouse monoclonal

Synonym(s):
Anti-Trans-Golgi network protein, 46-kD
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2