Anti-Rabbit IgG (whole molecule)–Alkaline Phosphatase antibody produced in goat
Products
Product Category
Brand
Biological Source
Clonality
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing