Skip to Content
Merck
Amikacin disulfate salt potency: 674-786 μg per mg (as amikacin base)

Amikacin disulfate salt

Amikacin disulfate salt potency: 674-786 μg per mg (as amikacin base)
Empirical Formula (Hill Notation):
C22H43N5O13 · 2H2SO4
CAS Number:
Molecular Weight:
781.76
EC Number:

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Activity Spectrum
Quality Segment
Mode of Action
Showing 1-1 of 1
Compare
Product Number
Description
Pricing