
Aluminum L-lactate

Synonym(s):
L-Lactic acid aluminum salt
Linear Formula:
[CH3CH(OH)COO]3Al
CAS Number:
Molecular Weight:
294.19
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing