
3-Butynoic acid

Empirical Formula (Hill Notation):
C4H4O2
CAS Number:
Molecular Weight:
84.07
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing