Skip to Content
Merck
2-Deoxy-L-ribose ≥97.0% (TLC)

2-Deoxy-L-ribose

2-Deoxy-L-ribose ≥97.0% (TLC)
Synonym(s):
2-Deoxy-L-erythro-pentose
Empirical Formula (Hill Notation):
C5H10O4
CAS Number:
Molecular Weight:
134.13

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing