
1,5-Dicaffeoylquinic acid

Synonym(s):
(1S,3R,4R,5R)-1,3-Bis[3-(3,4-dihydroxyphenyl)acryloyloxy]-4,5-dihydroxycyclohexanecarboxylic acid, 1,3-Dicaffeoylquinic acid
Empirical Formula (Hill Notation):
C25H24O12
CAS Number:
Molecular Weight:
516.45
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing