Flow Cytometry Antibodies
Flow cytometry analysis is a technique commonly used to examine physical and chemical characteristics of cells and particles. Antibodies are used in flow cytometry to help characterize markers on the cell surface since they allow for the detection of specific antigens. These antibodies can be directly conjugated to a fluorophore (direct detection) or paired with a fluorescently labeled secondary antibody (indirect detection). By adding specific fluorophores to specific antibodies, cells can be sorted and analyzed simultaneously.
For successful flow cytometry assays and reproducible results, antibodies validated for use in flow cytometry and proper controls are essential. We offer high-quality flow cytometry validated primary antibodies, secondary antibodies, and isotype control antibodies to support all your flow cytometry needs.
Need Help Finding The Correct Antibody for Your Application?
Use our Antibody Explorer search tool to view and compare antibodies by clonality, application, species, reactivity, conjugate, host species, and form.
Not finding the right product?
Try our Product Selector Tool.
For Research Use Only. Not For Use In Diagnostic Procedures.
Unless otherwise stated in the Product(s) specifications, any Antibody product is sold for internal research use only and may not be used for any other purpose, which includes but is not limited to, any commercial, diagnostic, or therapeutic use. Our validation processes pertain only to research uses and do not confirm or assure that our antibodies can be used for any unauthorized uses as set forth herein.
Related Resources
- Article: Flow Cytometry Guide
Explore our flow cytometry guide to uncover flow cytometry basics, traditional flow cytometer components, key flow cytometry protocol steps, and proper controls.
- Article: Key Steps in Flow Cytometry Protocols
Learn key steps in flow cytometry protocols to make your next flow cytometry experiment run with ease.
- Article: Multicolor Flow Cytometry
Uncover tips for multicolor flow cytometry experiments, including the challenges in multicolor flow cytometry, steps for how to build a multicolor flow cytometry panel, tandem dyes, and tools for multicolor assay design.
- Article: How to Select Flow Cytometry Dyes
Flow cytometry dye selection tips match fluorophores to flow cytometer configurations, enhancing panel performance.
- Article: Flow Cytometry Troubleshooting Guide
Troubleshooting guide offers solutions for common flow cytometry problems, ensuring improved analysis performance.
- Article: How to Choose a Secondary Antibody
Review the key factors that should figure in your decision to choose a secondary antibody. Learn about species, subclass, isotype, label, and more.