Skip to Content
Merck
All Photos(1)

Documents

K562 Cell line human

89121407, human blood (chronic myelogenous leukemia), Lymphoblast

Synonym(s):

K-562 Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

K562 Cell line human, 89121407, from human blood(chronic myelogenous leukemia)

biological source

human blood (chronic myelogenous leukemia)

growth mode

Suspension

karyotype

2n = 46

morphology

Lymphoblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Caucasian chronic myelogenous leukaemia

Cell Line Description

Established from pleural effusion of 53 year old female with chronic myelogenous leukaemia in terminal blast crisis. Population highly undifferentiated and of the granulocytic series. Can be used as a highly sensitive target for the in vitro natural killer assay. Recent studies have shown the K562 blasts are multipotential, hematopoietic malignant cells that spontaneously differentiate into recognisable progenitors of the erythrocyte, granulocyte and monocytic series.

Application

K562 cell line has been used to study the protecting role of cluster of differentiation-39 (CD39) attenuation against DNA damage. It has also been used to study the relevance of a novel sorting approach for separating cells, called “Slice and Dice Sorting”.
Natural killer assay and tumourigenicity studies

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 9,10
D13S317: 8
D16S539: 11,12
D5S818: 11,12
D7S820: 9,11
THO1: 9.3
TPOX: 8,9
vWA: 16

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 100,000 to 1,000,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service