Skip to Content
Merck
All Photos(1)

Key Documents

MAB994-I

Sigma-Aldrich

Anti-Reovirus Type 3 sigma 1 hemagglutinin Antibody, clone 9BG5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

antibody form

purified antibody

clone

9BG5
monoclonal

purified by

using protein G

species reactivity

mouse, virus

concentration

(Please refer to lot specific datasheet.)

technique(s)

flow cytometry: suitable
immunofluorescence: suitable
radioimmunoassay: suitable

isotype

IgG2aκ

capacity

neutralization

General description

Clone 9BG5 detects hemaglutinin protein of type 3 reovirus. Specific for the sigma 1 hemagglutinin of Type 3 Reovirus. It neutralizes reovirus type 3 and inhibits the hemagglutinating activity of the virus.
Purified reovirus type 3 virons.

Application

Immunofluorescence Analysis: A representative lot detected Reovirus Type 3 sigma 1 hemagglutinin in Immunofluorescence applications (Barton, E.S., et. al. (2001). J Biol Chem. 276(3):2200-2211).

Radioimmunoassay Analysis: A representative lot detected Reovirus Type 3 sigma 1 hemagglutinin in Radioimmunoassay applications (William, W.V., et. al. (1988). Proc Natl Acad Sci U S A. 85(17):6488-92).

Flow Cytometry Analysis: A representative lot detected Reovirus Type 3 sigma 1 hemagglutinin in Flow Cytometry applications (Dietrich, M.H., et. al. (2017). J Virol. 91(4). pii: e01621-16).

Neutralizing Analysis: A representative lot neutralized Reovirus Type 3 in Neutralizing applications (William, W.V., et. al. (1988). Proc Natl Acad Sci USA. 85(17):6488-92).

Quality

Isotype testing: Identity Confirmation by Isotyping Test.

Isotyping Analysis: The identity of this monoclonal antibody is confirmed by isotyping test to be mouse IgG2a.

Physical form

Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Virus (Reovirus type 3).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service