ABE411-S
Anti-dimethyl Histone H3 (Arg26), Asymmetric, Trial Size Antibody
from rabbit, purified by affinity chromatography
Synonym(s):
Histone H3.1, Histone H3/a, Histone H3/b, Histone H3/c, Histone H3/d, Histone H3/f, Histone H3/h, Histone H3/i, Histone H3/j, Histone H3/k, Histone H3/l
About This Item
biological source
rabbit
Quality Level
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
purified by
affinity chromatography
species reactivity
human
technique(s)
dot blot: suitable
immunocytochemistry: suitable
western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
dimethylation (Arg26)
Gene Information
human ... HIST1H3F(8968)
General description
Specificity
Immunogen
Application
Dot Blot (Specificity) Analysis: 0.5 µg/mL from a representative lot detected dimethyl Histone H3 (Arg26), Asymmetric in Absurance Histone H3 Antibody Specificity Array (Cat. No. 16-667) and Absurance Histone H2A, H2B, H4 Antibody Specificity Array (Cat. No. 16-665).
DyLight is a registered trademark of Thermo Fisher Scientific.
Epigenetics & Nuclear Function
Histones
Quality
Western Blotting Analysis: 1 µg/mL of this antibody detects dimethyl Histone H3 (Arg26), Asymmetric in 10 µg of CARM1 treated HeLa acid extract.
Target description
Physical form
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service