Skip to Content
Merck
All Photos(1)

Key Documents

40-014

Millipore

FLEXMAP 3D System®

The FLEXMAP 3D system makes it possible to multiplex up to 500 analytes through the combination of differentially dyed fluorescent microsphere sets & an innovative instrument design.

Synonym(s):

3D System, FLEXMAP 3D

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116133
eCl@ss:
32161000
NACRES:
NB.51

Quality Level

manufacturer/tradename

Milliplex®

technique(s)

multiplexing: suitable

detection method

fluorometric

General description

The FLEXMAP 3D system® makes it possible to multiplex up to 500 analytes through the combination of differentially dyed fluorescent microsphere sets and an innovative instrument design. Features such as automated probe height adjustment, simplified routine maintenance operations and an intuitive software interface make FLEXMAP 3D a flexible and user-friendly multiplexing platform capable of performing a variety of protein and nucleic acid applications. The new xPONENT 4.0 software controls the FLEXMAP 3D system® and offers interfacing options for laboratory information systems (LIS) as well as automation.

Application

The FLEXMAP 3D system makes it possible to multiplex up to 500 analytes through the combination of differentially dyed fluorescent microsphere sets & an innovative instrument design.

Legal Information

FLEXMAP 3D system is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service