Skip to Content
Merck
All Photos(1)

Documents

615366

Sigma-Aldrich

Vitamin E acetate-(trimethyl-d9)

98 atom % D, 98% (CP)

Synonym(s):

2RS,4′RS,8′RS-O-Acetyl-α-tocopherol-d9 (5,7,8-tri(methyl-d3))

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31D9H43O3
Molecular Weight:
481.80
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.12

description

8 isomer mix

Quality Level

isotopic purity

98 atom % D

Assay

98% (CP)

form

liquid

technique(s)

mass spectrometry (MS): suitable

bp

224 °C/0.3 mmHg (lit.)

mass shift

M+9

SMILES string

[2H]C([2H])([2H])c1c2CC[C@@](C)(CCCC(C)CCCC(C)CCCC(C)C)Oc2c(c(c1OC(C)=O)C([2H])([2H])[2H])C([2H])([2H])[2H]

InChI

1S/C31H52O3/c1-21(2)13-10-14-22(3)15-11-16-23(4)17-12-19-31(9)20-18-28-26(7)29(33-27(8)32)24(5)25(6)30(28)34-31/h21-23H,10-20H2,1-9H3/t22?,23?,31-/m1/s1/i5D3,6D3,7D3

InChI key

ZAKOWWREFLAJOT-JTMQEPRKSA-N

Application

Vitamin E acetate-(trimethyl-d9) or vitamin E acetate D9 can be used as an internal standard to study dietary intake, bioavailability, identification and concentration within the complex biological samples.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service