Skip to Content
Merck
All Photos(1)

Key Documents

374199

Sigma-Aldrich

1-Octanesulfonyl chloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7SO2Cl
CAS Number:
Molecular Weight:
212.74
Beilstein:
1763921
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.460 (lit.)

bp

75 °C/0.03 mmHg (lit.)

density

1.087 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCS(Cl)(=O)=O

InChI

1S/C8H17ClO2S/c1-2-3-4-5-6-7-8-12(9,10)11/h2-8H2,1H3

InChI key

WIVNTNLDTMNDNO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-Octanesulfonyl chloride is suitable for use in the preparation of 1-alkyl-3-methylimidazolium alkylsulfonates (ionic liquids).

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marijana Blesic et al.
Physical chemistry chemical physics : PCCP, 11(21), 4260-4268 (2009-05-22)
Anionic and cationic alkyl-chain effects on the self-aggregation of both neat and aqueous solutions of 1-alkyl-3-methylimidazolium alkylsulfonate salts ([C(n)H(2n+1)mim][C(m)H(2m+1)SO(3)]; n = 8, 10 or 12; m = 1 and n = 4 or 8; m = 4 or 8) have

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service