Merck
  • C 3-symmetric opioid scaffolds are pH-responsive DNA condensation agents.
C 3-symmetric opioid scaffolds are pH-responsive DNA condensation agents.
Nucleic acids research (2016-12-03)
Natasha McStay, Zara Molphy, Alan Coughlan, Attilio Cafolla, Vickie McKee, Nicholas Gathergood, Andrew Kellett
ABSTRACT
Herein we report the synthesis of tripodal C
Sigma-Aldrich
Poly(deoxyadenylic-thymidylic) acid sodium salt, double-stranded alternating copolymer
Sigma-Aldrich
Deoxyribonucleic acid sodium salt from salmon testes
Sigma-Aldrich
Poly(deoxyguanylic-deoxycytidylic) acid sodium salt, double stranded, alternating copolymer