Merck
  • Culture-negative pericarditis caused by Neisseria meningitidis serogroup C.
Culture-negative pericarditis caused by Neisseria meningitidis serogroup C.
Journal of clinical microbiology (2004-02-10)
Kaoutar Moumile, Etienne Carbonnelle, Philippe Dessemme, Daniel Tamisier, Franck Iserin, Véronique Houdouin, Xavier Nassif, Patrick Berche
ABSTRACT
We describe a case of primary purulent culture-negative pericarditis caused by Neisseria meningitidis serogroup C occurring in an 8-month-old previously healthy boy, which was detected in pericardial fluid by broad-spectrum PCR amplification.
Sigma-Aldrich
Polystyrene-block-poly(acrylic acid), DDMAT terminated, PS:PAA 3,000:5,000, PDI ≤1.1
Sigma-Aldrich
Poly(styrene)-block-poly(acrylic acid), average Mn 38,000, 20 wt. % PAA, PDI ≤1.3
Sigma-Aldrich
Poly(styrene)-block-poly(acrylic acid), PS:PAA 30,000:2,000, PDI ≤1.1
Sigma-Aldrich
Polystyrene-block-poly(acrylic acid)