Merck
  • The paracaspase MALT1 cleaves HOIL1 reducing linear ubiquitination by LUBAC to dampen lymphocyte NF-κB signalling.
The paracaspase MALT1 cleaves HOIL1 reducing linear ubiquitination by LUBAC to dampen lymphocyte NF-κB signalling.
Nature communications (2015-11-04)
Theo Klein, Shan-Yu Fung, Florian Renner, Michael A Blank, Antoine Dufour, Sohyeong Kang, Madison Bolger-Munro, Joshua M Scurll, John J Priatel, Patrick Schweigler, Samu Melkko, Michael R Gold, Rosa I Viner, Catherine H Régnier, Stuart E Turvey, Christopher M Overall
ABSTRACT
Antigen receptor signalling activates the canonical NF-κB pathway via the CARD11/BCL10/MALT1 (CBM) signalosome involving key, yet ill-defined roles for linear ubiquitination. The paracaspase MALT1 cleaves and removes negative checkpoint proteins, amplifying lymphocyte responses in NF-κB activation and in B-cell lymphoma subtypes. To identify new human MALT1 substrates, we compare B cells from the only known living MALT1(mut/mut) patient with healthy MALT1(+/mut) family members using 10-plex Tandem Mass Tag TAILS N-terminal peptide proteomics. We identify HOIL1 of the linear ubiquitin chain assembly complex as a novel MALT1 substrate. We show linear ubiquitination at B-cell receptor microclusters and signalosomes. Late in the NF-κB activation cycle HOIL1 cleavage transiently reduces linear ubiquitination, including of NEMO and RIP1, dampening NF-κB activation and preventing reactivation. By regulating linear ubiquitination, MALT1 is both a positive and negative pleiotropic regulator of the human canonical NF-κB pathway-first promoting activation via the CBM--then triggering HOIL1-dependent negative-feedback termination, preventing reactivation.
Sigma-Aldrich
MISSION® esiRNA, targeting human RBCK1
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-14-3-3 ε Antibody, CT, serum, Chemicon®
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-RBCK1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution