Merck
  • Graft copolymers that exhibit temperature-induced phase transitions over a wide range of pH.
Graft copolymers that exhibit temperature-induced phase transitions over a wide range of pH.
Nature (1995-01-05)
G Chen, A S Hoffman
ABSTRACT
There are many potential applications of 'intelligent' aqueous polymer systems in medicine, biotechnology, industry and in environmental problems. Many of these polymer systems undergo reversible phase transitions--for example, abrupt changes in volume--in response to external stimuli such as temperature, pH or the nature of the solvent. Most of the polymers studied previously are responsive to only one kind of stimulus. But for some applications, independent responsiveness to several factors, such as temperature and pH, may be required. Here we describe a polymer that undergoes marked solubility changes in water in response to temperature and/or pH changes. The polymer is prepared by grafting temperature-sensitive side chains onto a pH-sensitive backbone. We also find that block copolymers, in which the temperature- and pH-sensitive units alternate along the chain, show similar behaviour.
Sigma-Aldrich
Poly(N-isopropylacrylamide), carboxylic acid terminated, average Mn 10,000
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Poly(N-isopropylacrylamide-co-butylacrylate), butylacrylate 12 mol %, average Mn 30,000
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Poly(N-isopropyl acrylamide), NHS ester end functionalized, average Mn 5,000