Merck
  • Ophiopogonin D maintains Ca2+ homeostasis in rat cardiomyocytes in vitro by upregulating CYP2J3/EETs and suppressing ER stress.
Ophiopogonin D maintains Ca2+ homeostasis in rat cardiomyocytes in vitro by upregulating CYP2J3/EETs and suppressing ER stress.
Acta pharmacologica Sinica (2016-02-04)
Wen-ting You, Tao Zhou, Zeng-chun Ma, Qian-de Liang, Cheng-rong Xiao, Xiang-lin Tang, Hong-ling Tan, Bo-li Zhang, Yu-guang Wang, Yue Gao
ABSTRACT
CYP2J3 in myocardium metabolizes arachidonic acid to 4 regioisomeric epoxyeicosatrienoic acids (EETs), which have diverse biological activities in rat heart. In this study we examined whether CYP2J3 was involved in cardioprotective effects of ophiopogonin D (OPD), a steroidal glycoside isolated from Chinese herb Radix ophiopogonis. Rat cardiomyoblast cell line (H9c2 cells) was tested. Intracellular Ca(2+) concentrations ([Ca(2+)]i) were measured using Fluo-4/AM. The expression of calcium-regulating molecules and ER stress signaling molecules was measured with qRT-PCR and Western blot analyses. Cell apoptosis was quantified with Hoechst 33258 staining and TUNEL assay. The level of 14,15-DHET, a stable metabolite of 14,15-EET, was assessed with ELISA. Angiotensin II (10(-6) mol/L) significantly decreased the expression of calcium-regulating molecules (SERCA2a, PLB, RyR2 and FKBP12.6), and elevated [Ca(2+)]i in H9c2 cells. Furthermore, angiotensin II markedly increased the expression of ER stress signaling molecules (GRP78, CHOP, p-JNK and cleaved caspase-12) and ER stress-mediated apoptosis. OPD (100, 250 and 500 nmol/L) dose-dependently increased CYP2J3 expression and 14,15-DHET levels in normal H9c2 cells. Pretreatment of H9c2 cells with OPD suppressed angiotensin II-induced abnormalities in Ca(2+) homeostasis, ER stress responses and apoptosis. Overexpression of CYP2J3 or addition of exogenous 14,15-EET also prevented angiotensin II-induced abnormalities in Ca(2+) homeostasis, whereas transfection with CYP2J3 siRNA diminished the effects of OPD on Ca(2+) homeostasis. Furthermore, the intracellular Ca(2+) chelator BAPTA suppressed angiotensin II-induced ER stress responses and apoptosis in H9c2 cells. OPD is a novel CYP2J3 inducer that may offer a therapeutic benefit in treatment of cardiovascular diseases related to disturbance of Ca(2+) homeostasis and ER stress.
Sigma-Aldrich
6-(2-Propargyloxyphenyl)hexanoic acid, ≥98%, solid
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film
Sigma-Aldrich
11,12-Epoxy-(5Z,8Z,14Z)-eicosatrienoic acid, ~100 μg/mL in ethanol, ≥95%