Merck
  • Simultaneous Determination of Nitrofuran Metabolites and Chloramphenicol in Shrimp with a Single Extraction and LC-MS/MS Analysis.
Simultaneous Determination of Nitrofuran Metabolites and Chloramphenicol in Shrimp with a Single Extraction and LC-MS/MS Analysis.
Journal of AOAC International (2015-05-31)
Aref El-Demerdash, Fenhong Song, Robin K Reel, Judy Hillegas, Robert E Smith
ABSTRACT
A method has been developed to quantify the nitrofuran metabolites 3-amino-5-morphorinomethyl-1,3-oxazolidinone, 3-amino-oxazolidinone, 1-aminohydantoin, and semicarbazide, as well as chloramphenicol in shrimp with a single extraction procedure followed by LC-MS/MS analysis. Dynamic selected reaction monitoring with positive and negative ionization mode switching was used. The method LODs were 0.5 ng/g for the nitrofuran metabolites and 0.3 ng/g for chloramphenicol.
Product Description
Supelco
2-NP-AHD-13C3, VETRANAL®, analytical standard
Sigma-Aldrich
Chloramphenicol, ≥98% (HPLC)
Sigma-Aldrich
Chloramphenicol, meets USP testing specifications
Sigma-Aldrich
Chloramphenicol, BioReagent, suitable for plant cell culture