Merck
  • Loss of MUC2 expression correlates with progression along the adenoma-carcinoma sequence pathway as well as de novo carcinogenesis in the colon.
Loss of MUC2 expression correlates with progression along the adenoma-carcinoma sequence pathway as well as de novo carcinogenesis in the colon.
Histology and histopathology (2006-12-14)
T Mizoshita, T Tsukamoto, K I Inada, N Hirano, M Tajika, T Nakamura, H Ban, M Tatematsu
ABSTRACT
We have previously demonstrated links between clinicopathological findings and phenotypes using several gastric and intestinal phenotypic markers in stomach and pancreatic cancers. However, the clinicopathological significance of the phenotype and Cdx2 expression has hitherto remained unclear in colorectal carcinogenesis. We examined the correlation between gastric and intestinal phenotypic expression in 91 primary early carcinomas of the colon. MUC2 expression demonstrated a significant decrease from tubular/tubulovillous adenomas with moderate atypia, through intramucosal carcinomas, to cancers with submucosal invasion (P<0.0001). Intramucosal de novo carcinomas (flat type carcinomas without adenomatous components) exhibited a greater decrease of MUC2 than intramucosal lesions with adenomatous components. Expression of MUC5AC also decreased significantly with progression according to the tubular/tubulovillous adenoma-carcinoma sequence, carcinomas with villous adenomatous components having a higher level compared with their tubular adenomatous counterparts, suggesting differences in the pathway of malignant transformation. Cdx2 nuclear expression was maintained in all of the adenomas and early carcinomas examined. Our data suggest that the reduction of MUC2 expression may be associated with the occurrence and progression of colorectal carcinomas in both adenoma-carcinoma sequence pathway and de novo carcinogenesis. Tumor-suppressive effects of Cdx2 may be preserved during early stages of colorectal carcinogenesis.
Sigma-Aldrich
MUC2 (MRQ-18) Mouse Monoclonal Antibody
Sigma-Aldrich
MUC1 (MRQ-17) Mouse Monoclonal Antibody
Sigma-Aldrich
MUC6 (MRQ-20) Mouse Monoclonal Antibody