- Miniaturised isotachophoresis analysis.
Miniaturised isotachophoresis analysis.
Lab on a chip (2006-03-31)
Lin Chen, Jeff E Prest, Peter R Fielden, Nicholas J Goddard, Andreas Manz, Philip J R Day
PMID16572209
ABSTRACT
The application of miniaturized total analysis systems (microTAS) has seen rapid development over the past few years. Isotachophoresis (ITP) has been transferred into microchip format for both electrophoretic separation and pretreatment purposes, due to its advantageous features including separation parameters controlled by electrolyte composition and high sample load capacity. The primary focus of this concise review is to summarize the basic features of microchip based ITP and its applications to the analysis and pretreatment of ionic compounds and biomolecules that have arisen since 1998.