Merck
  • [Modification of DNA damage in rat hepatocytes following whole-body roentgen radiation using cysteamine and 8-bromocaffeine].
[Modification of DNA damage in rat hepatocytes following whole-body roentgen radiation using cysteamine and 8-bromocaffeine].
Radiobiologiia (1983-09-01)
I S Smirnova, A V Suslov, L A Noskin
ABSTRACT
Using the elastoviscosimetric method it was shown that the level of residual DNA double-strand breaks in rat hepatocytes depended on conditions of total-body X-irradiation: it increased in the presence of a sensitizer, 8-bromocaffeine and decreased in the presence of a protector, cysteamine. This correlation confirms a major role of fixed DNA double-strand breaks in the formation of a lethal effect in a cell.
Sigma-Aldrich
8-Bromocaffeine, 98%