Merck
  • New V(IV)-based metal-organic framework having framework flexibility and high CO2 adsorption capacity.
New V(IV)-based metal-organic framework having framework flexibility and high CO2 adsorption capacity.
Inorganic chemistry (2012-12-22)
Ying-Ya Liu, Sarah Couck, Matthias Vandichel, Maciej Grzywa, Karen Leus, Shyam Biswas, Dirk Volkmer, Jorge Gascon, Freek Kapteijn, Joeri F M Denayer, Michel Waroquier, Veronique Van Speybroeck, Pascal Van Der Voort
ABSTRACT
A vanadium based metal-organic framework (MOF), VO(BPDC) (BPDC(2-) = biphenyl-4,4'-dicarboxylate), adopting an expanded MIL-47 structure type, has been synthesized via solvothermal and microwave methods. Its structural and gas/vapor sorption properties have been studied. This compound displays a distinct breathing effect toward certain adsorptives at workable temperatures. The sorption isotherms of CO(2) and CH(4) indicate a different sorption behavior at specific temperatures. In situ synchrotron X-ray powder diffraction measurements and molecular simulations have been utilized to characterize the structural transition. The experimental measurements clearly suggest the existence of both narrow pore and large pore forms. A free energy profile along the pore angle was computationally determined for the empty host framework. Apart from a regular large pore and a regular narrow pore form, an overstretched narrow pore form has also been found. Additionally, a variety of spectroscopic techniques combined with N(2) adsorption/desorption isotherms measured at 77 K demonstrate that the existence of the mixed oxidation states V(III)/V(IV) in the titled MOF structure compared to pure V(IV) increases the difficulty in triggering the flexibility of the framework.
Sigma-Aldrich
Vanadium, foil, thickness 1.0 mm, 99.7%
Sigma-Aldrich
Vanadium, foil, thickness 0.127 mm, 99.7% trace metals basis
Sigma-Aldrich
Vanadium, powder, −325 mesh, 99.5% trace metals basis
Sigma-Aldrich
Vanadium, foil, thickness 0.25 mm, 99.7% trace metals basis