Merck
  • The effect of cooking and washing rice on the bio-accessibility of As, Cu, Fe, V and Zn using an on-line continuous leaching method.
The effect of cooking and washing rice on the bio-accessibility of As, Cu, Fe, V and Zn using an on-line continuous leaching method.
Analytica chimica acta (2012-12-19)
Nolan S Horner, Diane Beauchemin
ABSTRACT
A previously developed method based on continuous on-line leaching with artificial gastro-intestinal fluids was used to determine the bio-accessible fraction of As, Cu, Fe, V and Zn in brown and white rice from California by inductively coupled mass spectrometry (ICP-MS). Saliva generally accounted for the largest percentage of total element leached in comparison to gastric and intestinal juices. Arsenic speciation analysis was performed on the saliva and gastric juice leachates using ion exchange chromatography coupled to ICP-MS. The four most toxic species of As (As(III), monomethylarsonic acid (MMA), dimethylarsinic acid (DMA) and As(V)), as well as Cl(-) in the gastric juice leachate, were successfully separated within 5.5min using a simple nitric acid gradient. While cooking rice had relatively little effect on total bio-accessibility, a change in species from As(V) and DMA to As(III) was observed for both types of rice. On the other hand, washing the rice with doubly deionized water prior to cooking removed a large percentage of the total bio-accessible fraction of As, Cu, Fe, V and Zn.
Sigma-Aldrich
Vanadium, foil, thickness 0.127 mm, 99.7% trace metals basis
Sigma-Aldrich
Vanadium, foil, thickness 1.0 mm, 99.7%
Sigma-Aldrich
Vanadium, foil, thickness 0.25 mm, 99.7% trace metals basis
Sigma-Aldrich
Vanadium, powder, −325 mesh, 99.5% trace metals basis