Merck
Genome-scale promoter engineering by coselection MAGE.
Nature methods (2012-04-10)
Harris H Wang, Hwangbeom Kim, Le Cong, Jaehwan Jeong, Duhee Bang, George M Church
ABSTRACT
Multiplex automated genome engineering (MAGE) uses short oligonucleotides to scarlessly modify genomes; however, insertions >10 bases are still inefficient but can be improved substantially by selection of highly modified chromosomes. Here we describe 'coselection' MAGE (CoS-MAGE) to optimize biosynthesis of aromatic amino acid derivatives by combinatorially inserting multiple T7 promoters simultaneously into 12 genomic operons. Promoter libraries can be quickly generated to study gain-of-function epistatic interactions in gene networks.
Sigma-Aldrich
Indigo, synthetic, Dye content 95 %