Merck
  • Field enhancement sample stacking for analysis of organic acids in traditional Chinese medicine by capillary electrophoresis.
Field enhancement sample stacking for analysis of organic acids in traditional Chinese medicine by capillary electrophoresis.
Journal of chromatography. A (2012-03-03)
Qianqian Zhu, Xueqin Xu, Yuanyuan Huang, Liangjun Xu, Guonan Chen
ABSTRACT
A technique known as field enhancement sample stacking (FESS) and capillary electrophoresis (CE) separation has been developed to analyze and detect organic acids in the three traditional Chinese medicines (such as Portulaca oleracea L., Crataegus pinnatifida and Aloe vera L.). In FESS, a reverse electrode polarity-stacking mode (REPSM) was applied as on-line preconcentration strategy. Under the optimized condition, the baseline separation of eight organic acids (linolenic acid, lauric acid, p-coumaric acid, ascorbic acid, benzoic acid, caffeic acid, succinic acid and fumaric acid) could be achieved within 20 min. Validation parameters of this method (such as detection limits, linearity and precision) were also evaluated. The detection limits ranged from 0.4 to 60 ng/mL. The results indicated that the proposed method was effective for the separation of mixtures of organic acids. Satisfactory recoveries were also obtained in the analysis of these organic acids in the above traditional Chinese medicine samples.
Sigma-Aldrich
Borax Anhydrous, BioUltra, anhydrous, ≥99%
Sigma-Aldrich
Borax Anhydrous, puriss. p.a., anhydrous, ≥98.0% (T)
Sigma-Aldrich
Sodium tetraborate, 99.998% trace metals basis
Sigma-Aldrich
Sodium tetraborate, 99%