Merck
Genotoxicity potential of a new natural formicide.
Environmental science and pollution research international (2011-09-08)
Sylvie Cotelle, Renan C Testolin, Anne-Sophie Foltête, Georgiana Bossardi-Rissardi, Rosilene A Silveira, Claudemir M Radetski
ABSTRACT
Assessment of environmental impacts from pesticide utilization should include genotoxicity studies, where the possible effects of mutagenic/genotoxic substances on individuals are assessed. In this study, the genotoxicity profile of the new formicide Macex® was evaluated with two genotoxicity tests, namely, the micronucleus test with mouse bone marrow and Vicia faba, and a mutagenicity test using the Ames Salmonella assay. The bacterial reverse mutation test (Salmonella typhimurium strains TA97, TA98, TA100, TA102, and TA1535), the Vicia root tip and mouse micronucleus tests were conducted according to published protocols. In the range of the formicide Macex® concentrations tested from 0.06 to 1.0 g L⁻¹ (or mgkg⁻¹ in the mouse test), no genotoxicity was observed in the prokaryotic or eukaryotic test organisms. However, at Macex® concentrations of 0.5 g L⁻¹ and above a significant decrease in the mitotic index (P ≤ 0.05) in the V. faba was observed. Micronucleus formation was likewise increased in the test organism at concentrations starting at 2.0 g L⁻¹. These data allow us to classify this natural formicide preparation as a product with no geno-environmental-impact when applied at recommended concentrations.
Product Description
Supelco
Aroclor 1254 solution, certified reference material, 500 mg/kg in transformer oil
Supelco
Aroclor 1254 solution, certified reference material, 50 mg/kg in transformer oil
Supelco
Aroclor 1254, ampule of 50 mg, analytical standard
Supelco
Aroclor 1254 solution, certified reference material, 200 μg/mL in methanol
Supelco
Aroclor 1254 solution, certified reference material, 1000 μg/mL in methanol