Merck
  • A study on quantum chemical calculations of 3-, 4-nitrobenzaldehyde oximes.
A study on quantum chemical calculations of 3-, 4-nitrobenzaldehyde oximes.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2011-06-21)
Halil Gökce, Semiha Bahçeli
ABSTRACT
The molecular geometry, vibrational frequencies, 1H and 13C NMR chemical shifts, UV-vis spectra, HOMO-LUMO analyses, molecular electrostatic potentials (MEPs), , thermodynamic properties and atomic charges of 3- and 4-Nitrobenzaldehyde oxime (C7H6N2O3) molecules have been investigated by using Hartree-Fock (HF) and density functional theory (DFT/B3LYP) methods with the 6-311++G(d, p) basis set. The calculated optimized geometric parameters (bond lengths and bond angles), the vibrational frequencies calculated and 13C and 1H NMR chemical shifts values for the mentioned compounds are in a very good agreement with the experimental data. Furthermore, the highest occupied molecular orbital (HOMO) and the lowest unoccupied molecular orbital (LUMO) have been simulated and the transition states, energy band gaps and molecular electrostatic potential (MEP) maps for each oxime compound have been determined. Additionally, we also report the infrared intensities and Raman activities for the compounds under study.
Sigma-Aldrich
3-Nitrobenzaldehyde, ReagentPlus®, 99%
Sigma-Aldrich
4-Nitrobenzaldehyde, 98% (GC)
Supelco
4-Nitrobenzaldehyde, for spectrophotometric det. of amino sugars, ≥99.0%