Merck
  • Oriented 2D covalent organic framework thin films on single-layer graphene.
Oriented 2D covalent organic framework thin films on single-layer graphene.
Science (New York, N.Y.) (2011-04-09)
John W Colson, Arthur R Woll, Arnab Mukherjee, Mark P Levendorf, Eric L Spitler, Virgil B Shields, Michael G Spencer, Jiwoong Park, William R Dichtel
ABSTRACT
Covalent organic frameworks (COFs), in which molecular building blocks form robust microporous networks, are usually synthesized as insoluble and unprocessable powders. We have grown two-dimensional (2D) COF films on single-layer graphene (SLG) under operationally simple solvothermal conditions. The layered films stack normal to the SLG surface and show improved crystallinity compared with COF powders. We used SLG surfaces supported on copper, silicon carbide, and transparent fused silica (SiO(2)) substrates, enabling optical spectroscopy of COFs in transmission mode. Three chemically distinct COF films grown on SLG exhibit similar vertical alignment and long-range order, and two of these are of interest for organic electronic devices for which thin-film formation is a prerequisite for characterizing their optoelectronic properties.
Product Description
Supelco
Mesitylene, analytical standard
Sigma-Aldrich
Mesitylene, 98%
Sigma-Aldrich
Mesitylene, reagent grade, 97%
Supelco
1,3,5-Trimethylbenzene solution, certified reference material, 5000 μg/mL in methanol